Order Entry
Germany
ContactUsLinkComponent
(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
Catalog # APOSPC440001-250MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-bi-2-napthol
Catalog # APOSPC440001-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :710.508 g/mol
Storage Temperature:Ambient
CAS Number:849939-13-5
Formula:C₃₆H₁₈F₁₂O₂
MDL Number:MFCD08689862