

2,3,5,6-Tetrafluorophenyl trifluoroacetate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:142685-25-4
:
Formula:C₈HF₇O₂
MDL Number:MFCD07784266
:262.081 g/mol
UN:3272
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient