Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
APOSPC430603-1G
:
Apollo Scientific
:
1-(3-Ethoxy-4-fluorobenzyl)piperidin-4-amine
APOSPC430603-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
252.328 g/mol
:
Ambient
:
1206970-26-4
:
C₁₄H₂₁FN₂O
:
MFCD14585244