Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate
tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate
Catalog # APOSPC430544-100MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-(2-fluoro-6-hydroxyphenyl)piperidine-1-carboxylate
Catalog # APOSPC430544-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :295.349 g/mol
Storage Temperature:Ambient
CAS Number:1206970-27-5
Formula:C₁₆H₂₂FNO₃
MDL Number:MFCD14585455