Ethyl 1-benzyl-5,5-difluoro-4-hydroxypiperidine-3-carboxylate
Catalog # APOSPC430163-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.313 g/mol
Storage Temperature:Ambient
CAS Number:1356339-08-6
Formula:C₁₅H₁₉F₂NO₃
MDL Number:MFCD21601887