tert-Butyl 3,3-difluoropiperidin-4-ylcarbamate
Catalog # APOSPC430144-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:236.259 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1263180-22-8
Formula:C₁₀H₁₈F₂N₂O₂
MDL Number:MFCD18791569