Order Entry
Germany
ContactUsLinkComponent
 
4-Chloro-2-fluoro-N-methylaniline 95%
Catalog # APOSPC421216-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2-fluoro-N-methylaniline 95%
Catalog # APOSPC421216-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :159.589 g/mol
Storage Temperature:Ambient
CAS Number:894099-96-8
Formula:C₇H₇ClFN
MDL Number:MFCD11655731