Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-3-Fluorocyclobutan-1-amine hydrochloride
cis-3-Fluorocyclobutan-1-amine hydrochloride 95%
cis-3-Fluorocyclobutan-1-amine hydrochloride 95%
:
Apollo Scientific
cis-3-Fluorocyclobutan-1-amine hydrochloride 95%
:
Apollo Scientific
:
1408075-13-7
:
:
C₄H₈FN·ClH
:
MFCD23106124
:
125.572 g/mol
:
GHS07
:
Ambient