Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
APOSPC410347-1G
:
Apollo Scientific
:
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
APOSPC410347-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
245.176 g/mol
:
Ambient
:
1005630-54-5
:
C₁₀H₇F₄N₃
:
MFCD04967238