Order Entry
Germany
ContactUsLinkComponent
(2-Amino-4,5,6,7-tetrahydro-1-benzothiophen-3-yl)(4-fluorophenyl)methanone
(2-Amino-4,5,6,7-tetrahydro-1-benzothiophen-3-yl)(4-fluorophenyl)methanone
Catalog # APOSPC410103-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-Amino-4,5,6,7-tetrahydro-1-benzothiophen-3-yl)(4-fluorophenyl)methanone
Catalog # APOSPC410103-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.341 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₄FNOS
MDL Number:MFCD09859280