1-[(2,2,2-Trifluoroethoxy)methyl]-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410032-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.603 g/mol
Storage Temperature:Ambient
CAS Number:1185098-59-2
Formula:C₆H₈F₃N₃O·ClH
MDL Number:MFCD04969858