Order Entry
Germany
ContactUsLinkComponent
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410010-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410010-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.111 g/mol
Storage Temperature:Ambient
CAS Number:1147222-53-4
Formula:C₁₀H₉ClFN₃·ClH
MDL Number:MFCD04967257