Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
811799-54-9
:
Formula:
C₁₃H₁₆FNO₃
MDL Number:
MFCD20921817
:
253.269 g/mol
Storage Temperature:
Ambient