Order Entry
Germany
ContactUsLinkComponent
3-(3-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(3-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSPC409515-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSPC409515-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.242 g/mol
Storage Temperature:Ambient
CAS Number:916423-50-2
Formula:C₁₂H₁₂FN₃
MDL Number:MFCD09258792