Order Entry
Germany
ContactUsLinkComponent
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408415-500MG
Supplier:  Apollo Scientific
CAS Number:  
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408415-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :409.496 g/mol
Storage Temperature:Ambient
CAS Number:1420816-71-2
Formula:C₂₄H₂₈FN₃O₂
MDL Number:MFCD05662633