3-Chloro-2-((S)-pyrrolidin-3-yloxy)-5-trifluoromethyl-pyridine hydrochloride
Catalog # APOSPC408394-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:303.108 g/mol
Storage Temperature:Ambient
CAS Number:1417789-61-7
Formula:C₁₀H₁₀ClF₃N₂O·ClH
MDL Number:MFCD22631740