Order Entry
Germany
ContactUsLinkComponent
(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester
(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408382-500MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-3-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yloxy]-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408382-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :411.469 g/mol
Storage Temperature:Ambient
CAS Number:1354006-88-4
Formula:C₂₃H₂₆FN₃O₃
MDL Number:MFCD21099002