[1-(4-Fluoro-benzenesulphonyl)-piperidin-3-yl]-methyl-amine hydrochloride
Catalog # APOSPC408365-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:308.8 g/mol
Storage Temperature:Ambient
CAS Number:1353972-68-5
Formula:C₁₂H₁₇FN₂O₂S·ClH
MDL Number:MFCD21098933