4-(4-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408342-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:341.441 g/mol
Storage Temperature:Ambient
CAS Number:1353989-67-9
Formula:C₁₇H₂₄FNO₃S
MDL Number:MFCD21098479