(2-Fluoro-benzyl)-methyl-piperidin-4-yl-amine hydrochloride
Catalog # APOSPC408309-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:258.763 g/mol
Storage Temperature:Ambient
CAS Number:1261235-83-9
Formula:C₁₃H₁₉FN₂·ClH
MDL Number:MFCD18380005