Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-2-methyl-7-trifluoromethylquinoline
4-Hydroxy-2-methyl-7-trifluoromethylquinoline
4-Hydroxy-2-methyl-7-trifluoromethylquinoline
4-Hydroxy-2-methyl-7-trifluoromethylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
4-Hydroxy-2-methyl-7-trifluoromethylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
15912-66-0
:
Formula:
C₁₁H₈F₃NO
MDL Number:
MFCD00100261
:
227.183 g/mol
Storage Temperature:
Ambient