Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Fluoro-4-hydroxy-2-phenylquinoline
6-Fluoro-4-hydroxy-2-phenylquinoline
6-Fluoro-4-hydroxy-2-phenylquinoline
6-Fluoro-4-hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
6-Fluoro-4-hydroxy-2-phenylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
103914-44-9
:
Formula:
C₁₅H₁₀FNO
MDL Number:
MFCD11505230
:
239.244 g/mol
Storage Temperature:
Ambient