tert-Butyl 5-fluoro-3-iodo-1H-indole-1-carboxylate
Catalog # APOSPC402039-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:361.151 g/mol
Storage Temperature:Ambient
CAS Number:192189-13-2
Formula:C₁₃H₁₃FINO₂
MDL Number:MFCD13183351