tert-Butyl 6-fluoro-2,3-dihydro-1H-indole-1-carboxylate
Catalog # APOSPC402023-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.27 g/mol
Storage Temperature:Ambient
CAS Number:1065183-64-3
Formula:C₁₃H₁₆FNO₂
MDL Number:MFCD13183504