tert-Butyl 3-(bromomethyl)-5-fluoro-1H-indazole-1-carboxylate
Catalog # APOSPC402003-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:329.165 g/mol
Storage Temperature:Ambient
CAS Number:944904-75-0
Formula:C₁₃H₁₄BrFN₂O₂
MDL Number:MFCD10696845