Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-4-fluorobenzo[b]furan
7-Bromo-4-fluorobenzo[b]furan
7-Bromo-4-fluorobenzo[b]furan
7-Bromo-4-fluorobenzo[b]furan
Supplier:
Apollo Scientific
Certificates
Expand image
7-Bromo-4-fluorobenzo[b]furan
Supplier:
Apollo Scientific
About this item
CAS Number:
253429-31-1
:
Formula:
C₈H₄BrFO
MDL Number:
MFCD09056781
:
215.019 g/mol
Melting Pt:
35…36 °C
:
GHS07
Storage Temperature:
Ambient