
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxaldehyde
Catalog # APOSPC3657-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.174 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1015845-52-9
Formula:C₁₀H₇FN₂O
MDL Number:MFCD05864518