
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazole-4-carboxaldehyde
Catalog # APOSPC32577-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.181 g/mol
Melting Pt:113…115 °C
:GHS07
Storage Temperature:Ambient
CAS Number:306936-65-2
Formula:C₁₁H₇F₃N₂O
MDL Number:MFCD02677691