Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%
APOSPC3255-1G
:
Apollo Scientific
:
Ethyl 5-methyl-2-(trifluoromethyl)-3-furoate 97%
APOSPC3255-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
222.161 g/mol
:
55 °C (0.3 mm Hg)
:
Ambient
:
17515-73-0
:
C₉H₉F₃O₃
:
MFCD00275564