Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-2,4-difluoro-3-methylaniline
6-Chloro-2,4-difluoro-3-methylaniline 97%
6-Chloro-2,4-difluoro-3-methylaniline 97%
6-Chloro-2,4-difluoro-3-methylaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Chloro-2,4-difluoro-3-methylaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2300095-53-6
:
Formula:
C₇H₆ClF₂N
MDL Number:
MFCD32640069
:
177.579 g/mol
Storage Temperature:
Ambient