Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethoxy-3,5-difluorobenzenesulphonamide
2-Ethoxy-3,5-difluorobenzenesulphonamide 97%
2-Ethoxy-3,5-difluorobenzenesulphonamide 97%
2-Ethoxy-3,5-difluorobenzenesulphonamide 97%
Supplier:
Apollo Scientific
Certificates
Bild vergrößern
2-Ethoxy-3,5-difluorobenzenesulphonamide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1409618-37-6
:
Formula:
C₈H₉F₂NO₃S
MDL Number:
MFCD19066363
:
237.224 g/mol
Storage Temperature:
Ambient