Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Ethoxy-2,3,4-trifluorobenzoic acid
6-Ethoxy-2,3,4-trifluorobenzoic acid 97%
6-Ethoxy-2,3,4-trifluorobenzoic acid 97%
APOSPC303287-1G
:
Apollo Scientific
:
6-Ethoxy-2,3,4-trifluorobenzoic acid 97%
APOSPC303287-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
220.145 g/mol
:
Ambient
:
1980050-06-3
:
C₉H₇F₃O₃
:
MFCD29277475