Order Entry
Germany
ContactUsLinkComponent
3-Ethoxy-2,4-difluorobenzenesulphonyl chloride
3-Ethoxy-2,4-difluorobenzenesulphonyl chloride
Catalog # APOSPC303258-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Ethoxy-2,4-difluorobenzenesulphonyl chloride
Catalog # APOSPC303258-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.654 g/mol
Storage Temperature:Ambient
CAS Number:1706439-10-2
Formula:C₈H₇ClF₂O₃S
MDL Number:MFCD28054223