Order Entry
Germany
ContactUsLinkComponent
4-Ethoxy-2,3-difluorobenzenesulphonyl chloride
4-Ethoxy-2,3-difluorobenzenesulphonyl chloride
Catalog # APOSPC303251-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-2,3-difluorobenzenesulphonyl chloride
Catalog # APOSPC303251-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.654 g/mol
Storage Temperature:Ambient
CAS Number:293299-90-8
Formula:C₈H₇ClF₂O₃S
MDL Number:MFCD17282233