Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoro-5-methoxy-4-methylaniline
2,3-Difluoro-5-methoxy-4-methylaniline 97%
2,3-Difluoro-5-methoxy-4-methylaniline 97%
2,3-Difluoro-5-methoxy-4-methylaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Difluoro-5-methoxy-4-methylaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1706446-33-4
:
Formula:
C₈H₉F₂NO
MDL Number:
MFCD27986845
:
173.16 g/mol
Storage Temperature:
Ambient