
4-Ethoxy-2,3-difluorobenzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:181806-55-3
:
Formula:C₉H₉BrF₂O
MDL Number:MFCD09258705
:251.068 g/mol
Melting Pt:58…61 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05