Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ethoxy-3,5-difluorobenzoic acid
4-Ethoxy-3,5-difluorobenzoic acid 97%
4-Ethoxy-3,5-difluorobenzoic acid 97%
Irritant
4-Ethoxy-3,5-difluorobenzoic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Ethoxy-3,5-difluorobenzoic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017779-19-9
:
Formula:
C₉H₈F₂O₃
MDL Number:
MFCD09258683
:
202.155 g/mol
:
GHS07
Storage Temperature:
Ambient