Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4-ethoxy-2,5-difluorobenzoate
Ethyl 4-ethoxy-2,5-difluorobenzoate
Ethyl 4-ethoxy-2,5-difluorobenzoate
Irritant
Ethyl 4-ethoxy-2,5-difluorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl 4-ethoxy-2,5-difluorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1823562-23-7
:
Formula:
C₁₁H₁₂F₂O₃
MDL Number:
MFCD28148236
:
230.208 g/mol
:
GHS07
Storage Temperature:
Ambient