Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
Irritant
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1379366-69-4
:
Formula:
C₈H₇ClF₃NO
MDL Number:
MFCD17168286
:
225.595 g/mol
:
GHS07
Storage Temperature:
Ambient