Order Entry
Germany
ContactUsLinkComponent
2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol, tech
2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol, tech
Catalog # APOSPC28130-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)-7-methylimidazo[1,2-a]pyrimidin-5-ol, tech
Catalog # APOSPC28130-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.236 g/mol
Storage Temperature:Ambient
CAS Number:866050-49-9
Formula:C₁₃H₁₀FN₃O
MDL Number:MFCD04125870