Order Entry
Germany
ContactUsLinkComponent
7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine, tech
7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine, tech
Catalog # APOSPC28047-1G
Supplier:  Apollo Scientific
CAS Number:  
7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine, tech
Catalog # APOSPC28047-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :285.316 g/mol
Storage Temperature:Ambient
CAS Number:478039-30-4
Formula:C₁₆H₁₆FN₃O
MDL Number:MFCD02571885