

3-Bromo-2,4,5-trifluorobenzyl fluoride 95%
Catalog # APOSPC27919-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:242.996 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
:GHS08
CAS Number:2244088-42-2
Formula:C₇H₃BrF₄
MDL Number:MFCD31664904