(R)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
Catalog # APOSPC210124-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:277.306 g/mol
Storage Temperature:Ambient
CAS Number:1547490-91-4
Formula:C₁₂H₁₄F₃NOS
MDL Number:MFCD18781370