Order Entry
Germany
ContactUsLinkComponent
tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate
tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate
Catalog # APOSPC210096-500MG
Supplier:  Apollo Scientific
CAS Number:  
tert-butyl 3-[(4-fluorophenyl)methyl]-4-oxo-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate
Catalog # APOSPC210096-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :359.395 g/mol
Storage Temperature:Ambient
CAS Number:1781241-50-6
Formula:C₁₉H₂₂FN₃O₃
MDL Number:MFCD28805993