Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-3-methylaniline
2,4-Difluoro-3-methylaniline 96%
2,4-Difluoro-3-methylaniline 96%
:
Apollo Scientific
2,4-Difluoro-3-methylaniline 96%
:
Apollo Scientific
:
76350-70-4
:
:
C₇H₇F₂N
:
MFCD09261073
:
143.134 g/mol
:
Ambient
:
2811
:
6.1,III
:
GHS06