
5-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
Catalog # APOSPC201197-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:237.649 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1074764-69-4
Formula:C₁₀H₁₀F₃N·ClH
MDL Number:MFCD05861552