Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5,7-Trifluoroadamantane-1-carboxylic acid
3,5,7-Trifluoroadamantane-1-carboxylic acid 95+%
3,5,7-Trifluoroadamantane-1-carboxylic acid 95+%
3,5,7-Trifluoroadamantane-1-carboxylic acid 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5,7-Trifluoroadamantane-1-carboxylic acid 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
214557-89-8
:
Formula:
C₁₁H₁₃F₃O₂
MDL Number:
MFCD17215150
:
234.215 g/mol
Storage Temperature:
Ambient