Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-6-Fluoroindan-1-amine hydrochloride
(S)-6-Fluoroindan-1-amine hydrochloride 95+%
(S)-6-Fluoroindan-1-amine hydrochloride 95+%
(S)-6-Fluoroindan-1-amine hydrochloride 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
(S)-6-Fluoroindan-1-amine hydrochloride 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1391354-92-9
:
Formula:
C₉H₁₀FN·ClH
MDL Number:
MFCD16295142
:
187.642 g/mol
Storage Temperature:
Ambient