2-(3,5-Difluorophenyl)propan-2-amine hydrochloride 98%
Catalog # APOSPC201015-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.648 g/mol
Storage Temperature:Ambient
CAS Number:1063733-81-2
Formula:C₉H₁₁F₂N·ClH
MDL Number:MFCD19687116