Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine
6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine 97%
6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine 97%
6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1036762-04-5
:
Formula:
C₁₂H₈FN₃
MDL Number:
MFCD28405285
:
213.21 g/mol
Storage Temperature:
Ambient